Learn More
Aldevron Dnase I, 70ug
Catalog No. 503073881
Encompass_Preferred
70ug dispense of Dnase I. DNase I nonspecifically cleaves single- or double-stranded DNA, producing di-, tri- and oligonucleotide products. DNase I is free of detectable ribonuclease activity.
Catalog No. 50-307-3881 Supplier Aldevron Supplier No. 913670MCG
70ug dispense of Dnase I. DNase I nonspecifically cleaves single- or double-stranded DNA, producing di-, tri- and oligonucleotide products. DNase I is free of detectable ribonuclease activity.