Learn More
STEMCELL Technologies DNase I, ACF, Size: 25000 U
SDP
Supplier:  STEMCELL Technologies 07473
Encompass
Deoxyribonuclease I (DNase I), Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials and purified by chromatography. DNase I is an endonuclease consisting of a single glycosylated polypeptide chain with two disulfide bonds. DNase is often included in tissue dissociation protocols to digest DNA that has leaked into the dissociation medium as a result of cell damage. DNase I, ACF preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides in both single- and double-stranded DNA, yielding polynucleotides with 5’-phosphate and 3’-hydroxyl groups (Bernardi et al.). DNase I has been used for the digestion of human tissues such as microglia (Klegeris & McGeer), cartilage (Dunham & Koch), colon (Fukushima & Fiocchi), epithelium l (Fukushima & Fiocchi), liver (Vatakis et al.), lung (Fujino et al.), neural (Fuja et al.), and stem cells (Kusuma et al.).
Catalog No. 50-197-4417