Learn More
Gold Biotechnology Inc DNase I, Bovine Pancreas (Lysate Tested), >2000 Kunitz U/mg 100 mg
SDP
Catalog No. NC2942871
Encompass_Preferred
Bovine pancreatic deoxyribonuclease (DNase) is an endonuclease, which splits phosphodiester linkages preferentially adjacent to a pyrimidine nucleotide yielding polynucleotides with a free hydroxyl group at the 3' position and a phosphate group at the 5' position. The optimum pH is 7.8.
Catalog No. NC2942871 Supplier Gold Biotechnology Inc Supplier No. D301100
Bovine pancreatic deoxyribonuclease (DNase) is an endonuclease, which splits phosphodiester linkages preferentially adjacent to a pyrimidine nucleotide yielding polynucleotides with a free hydroxyl group at the 3' position and a phosphate group at the 5' position. The optimum pH is 7.8.