Learn More
Worthington Biochemical Corporation DNASE I DPFE 5MG
SDP
Catalog No. NC9095598
Encompass
Deoxyribonuclease I, Chromatographically purified, A lyophilized powder containing glycine as a stabilizer, Contains /=2000 Kunitz U/mg dry
Catalog No. NC9095598 Supplier Worthington Biochemical Corporation Supplier No. LS006330
Deoxyribonuclease I, Chromatographically purified, A lyophilized powder containing glycine as a stabilizer, Contains /=2000 Kunitz U/mg dry