Learn More
Sigma Aldrich Fine Chemicals Biosciences DNase I recombinant, 2 x 10000 units
Catalog No. NC1243893
Encompass
DNase I, recombinant, grade I is used for: Eliminating DNA during protein isolation procedures. Analysis of chromatin structure. Eliminating DNA during sample preparation.
Catalog No. NC1243893 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4536282001
DNase I, recombinant, grade I is used for: Eliminating DNA during protein isolation procedures. Analysis of chromatin structure. Eliminating DNA during sample preparation.