Learn More
New England Biolabs, Inc. DNase I (RNase-free) – 5000 units
SDP
Supplier:  New England Biolabs, Inc. M0303L
Encompass
DNase I, (RNase-free) is an endonuclease that nonspecifically cleaves DNA to release di-, tri- and oligonucleotide products with 5' -phosphorylated and 3' -hydroxylated ends. DNase I acts on single- and double-stranded DNA, chromatin and RNA:DNA hybrids.
Catalog No. 50-995-226