Learn More
Apexbio Technology LLC DNase I (RNase-free) 10000U
SDP
Catalog No. 502240956
Encompass_Preferred
DNase I (RNase-free) is an endodeoxyribonuclease capable of hydrolyzing single-stranded or double-stranded DNA producing dinucleotide trinucleotide and oligonucleotide fragments with 3 -OH and 5 -phosphate ends The activity of DNase I (RNase-free) depends on Ca2 and can be activated by Mg2 or Mn2 In the presence of Mg2 DNase I (RNase-free) randomly cleaves double-stranded DNA at any site in the presence of Mn2 it simultaneously recognizes both DNA strands and cleaves at nearly identical sites DNase I (RNase-free) acts on single-stranded DNA double-stranded DNA chromatin and RNA DNA hybrids In molecular biology experiments DNase I (RNase-free) is suitable for the removal of DNA in RNA extraction in vitro transcription and RT-PCR applications
This item is not returnable. View return policy
Catalog No. 50-224-0956 Supplier Apexbio Technology LLC Supplier No. K108810000
DNase I (RNase-free) is an endodeoxyribonuclease capable of hydrolyzing single-stranded or double-stranded DNA producing dinucleotide trinucleotide and oligonucleotide fragments with 3 -OH and 5 -phosphate ends The activity of DNase I (RNase-free) depends on Ca2 and can be activated by Mg2 or Mn2 In the presence of Mg2 DNase I (RNase-free) randomly cleaves double-stranded DNA at any site in the presence of Mn2 it simultaneously recognizes both DNA strands and cleaves at nearly identical sites DNase I (RNase-free) acts on single-stranded DNA double-stranded DNA chromatin and RNA DNA hybrids In molecular biology experiments DNase I (RNase-free) is suitable for the removal of DNA in RNA extraction in vitro transcription and RT-PCR applications