Learn More
Apexbio Technology LLC DNase I (RNase-free),1000U
SDP
Catalog No. 502240954
Encompass_Preferred
DNase I (RNase-free) is an endonuclease that digests single- or double- stranded DNA, generating dinucleotide, trinucleotide, and oligonucleotide products with 5´-phosphorylated and 3´-hydroxylated ends. The activity of DNase I (RNase-free) dependent on Ca2+ and can be activated by Mg2+ or Mn2+. In the presence of Mg2+, DNase I (RNase-free) can randomly cleaves arbitrary sites of double-stranded DNA; in the presence of Mn2+, DNase I (RNase-free) can simultaneously recognize both strands of DNA and cleaves at nearly same sites. DNase I (RNase-free) can act on single stranded DNA, double stranded DNA, chromatin, and RNA: DNA hybrid strands. DNase I (RNase-free) is suitable for RNA extraction, in vitro transcription, and removal of DNA for RT-PCR experiments.Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0954 Supplier Apexbio Technology LLC Supplier No. K10881000
DNase I (RNase-free) is an endonuclease that digests single- or double- stranded DNA, generating dinucleotide, trinucleotide, and oligonucleotide products with 5´-phosphorylated and 3´-hydroxylated ends. The activity of DNase I (RNase-free) dependent on Ca2+ and can be activated by Mg2+ or Mn2+. In the presence of Mg2+, DNase I (RNase-free) can randomly cleaves arbitrary sites of double-stranded DNA; in the presence of Mn2+, DNase I (RNase-free) can simultaneously recognize both strands of DNA and cleaves at nearly same sites. DNase I (RNase-free) can act on single stranded DNA, double stranded DNA, chromatin, and RNA: DNA hybrid strands. DNase I (RNase-free) is suitable for RNA extraction, in vitro transcription, and removal of DNA for RT-PCR experiments.Other sizes are also available. Please inqury us for quote.