Learn More
Zymo Research Corporation DNase I Set (250 U) w/ DNA Digestion Buffer (4 ml)
SDP
Catalog No. NC0847937
Encompass_Preferred
It is typically used for selectively degrading DNA in the presence of RNA. This DNase is suited for applications such as nick translation, production of random fragments, cleavage of genomic DNA for footprinting, removal of DNA template after in vitro transcription, and removal of DNA from RNA samples prior to applications such as RT-PCR. It is compatible with all of our RNA kits featuring in-column DNase digestion. The DNase I Set features lyophilized enzyme and DNA Digestion Buffer.
Catalog No. NC0847937 Supplier Zymo Research Corporation Supplier No. E1010
It is typically used for selectively degrading DNA in the presence of RNA. This DNase is suited for applications such as nick translation, production of random fragments, cleavage of genomic DNA for footprinting, removal of DNA template after in vitro transcription, and removal of DNA from RNA samples prior to applications such as RT-PCR. It is compatible with all of our RNA kits featuring in-column DNase digestion. The DNase I Set features lyophilized enzyme and DNA Digestion Buffer.