Learn More
STEMCELL Technologies DNase I Solution (1 mg/mL), Size: 10 mL
SDP
Catalog No. NC2030908
Encompass
Deoxyribonuclease (DNase) I Solution (1 mg/mL) is useful to reduce or prevent the clumping of concentrated and/or cryopreserved cell suspensions following thawing. The activity range is ≥ 2000 units per mg of protein.
Catalog No. NC2030908 Supplier STEMCELL Technologies Supplier No. 1000762
Deoxyribonuclease (DNase) I Solution (1 mg/mL) is useful to reduce or prevent the clumping of concentrated and/or cryopreserved cell suspensions following thawing. The activity range is ≥ 2000 units per mg of protein.