Learn More
Active Motif DNase I Treatment Kit
SDP
Catalog No. NC1841578
Encompass
The DNase I Treatment Kit efficiently removes genomic DNA from RNA samples. It utilizes an endonuclease that non-specifically cleaves ssDNA, dsDNA and the DNA strand of RNA:DNA hybrids to improve the purity of your RNA samples. Following DNase I treatment, RNA samples are purified with easy-to-use spin columns. The RNA is then suitable for use in downstream applications such as RT-qPCR and RNA-seq. For more complete information, please visit the Active Motif website.
Catalog No. NC1841578 Supplier Active Motif Supplier No. 25502
The DNase I Treatment Kit efficiently removes genomic DNA from RNA samples. It utilizes an endonuclease that non-specifically cleaves ssDNA, dsDNA and the DNA strand of RNA:DNA hybrids to improve the purity of your RNA samples. Following DNase I treatment, RNA samples are purified with easy-to-use spin columns. The RNA is then suitable for use in downstream applications such as RT-qPCR and RNA-seq. For more complete information, please visit the Active Motif website.