Learn More
Bioworld DNase I (Bovine Pancreas), 50 KU
Catalog No. 50198970
Encompass_Preferred
DNase I DNase I is an endonuclease derived from bovine pancreas that will degrade double-stranded DNA in the presence of divalent cations, producing 3'-OH oligonucleotides. Mg+2 in the reaction solution causes the enzyme to produce nicks in double-stranded DNA, while in the presence on Mn+2, DNase I cleaves both strands of the DNA. DNase I is useful in nick translation for introducing single-stranded nicks that serve as primer sites for initiation of DNA synthesis and for cloning random DNA fragments by cleaving double-stranded DNA. It is also used in purifying RNA from cells or transcription mixtures. Specifications:CAS: 9003-98-9EC: 232-667-0Source: Bovine pancreasActivity: ≥10000 U/mg.Ultra Pure GradeOur DNase I is guaranteed free of RNase contaminants.
Catalog No. 50-198-970 Supplier Bioworld Supplier No. 215600082
DNase I DNase I is an endonuclease derived from bovine pancreas that will degrade double-stranded DNA in the presence of divalent cations, producing 3'-OH oligonucleotides. Mg+2 in the reaction solution causes the enzyme to produce nicks in double-stranded DNA, while in the presence on Mn+2, DNase I cleaves both strands of the DNA. DNase I is useful in nick translation for introducing single-stranded nicks that serve as primer sites for initiation of DNA synthesis and for cloning random DNA fragments by cleaving double-stranded DNA. It is also used in purifying RNA from cells or transcription mixtures. Specifications:CAS: 9003-98-9EC: 232-667-0Source: Bovine pancreasActivity: ≥10000 U/mg.Ultra Pure GradeOur DNase I is guaranteed free of RNase contaminants.