Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNase-, RNase-Free Water, MP Biomedicals™

Catalog No. 04821932 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-821-932 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-821-932 Supplier MP Biomedicals, Inc Supplier No. 04821932
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for the investigation of nucleic acid and protein synthesis.

  • Grade: DNase- RNase-free
  • Key applications: Investigation of nucleic acid and protein synthesis
  • Clear, colorless, liquid
  • 0.2 μ filtered (sterile)
TRUSTED_SUSTAINABILITY

Specifications

Color Clear, Colorless
CAS 7732-18-5
Boiling Point 100°C
DNase DNase-free
Molecular Formula H2O
MDL Number MFCD00011332
UN Number 0000
Merck Index 13.10098
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade Molecular Biology
Density 1 g/cm3 (4°C)
Quantity 500 mL
Refractive Index n20/D 1.34(Lit.)
Show More Show Less
EINECSNumber 231-791-2
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.