Learn More
Aviva Systems Biology Corp DNASE1 Antibody - N-terminal region (ARP46588_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46588_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNASE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DNASE1. Alternate names: DKFZp686H0155, DNL1, DRNI, FLJ38093, FLJ44902ApplicationWB. Species reactivity: Cow, Human, Rat
Catalog No. 50-014-59356