Learn More
Aviva Systems Biology Corp DNASE2B Antibody - middle region (ARP42596_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42596_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNASE2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DNASE2B. Alternate names: DLADApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-58278