Learn More
Molecular Research Center DNAzol, 200 mL
SDP
Catalog No. NC1213858
Encompass
DNAzol is for use with tissues, cells and liquid samples. 1.0 ml of DNAzol isolates DNA from 25 - 50 mg tissue or from cell pellets containing 1 - 3 x 10^7 cells.
Catalog No. NC1213858 Supplier Molecular Research Center Supplier No. DN 127200ML
DNAzol is for use with tissues, cells and liquid samples. 1.0 ml of DNAzol isolates DNA from 25 - 50 mg tissue or from cell pellets containing 1 - 3 x 10^7 cells.