Learn More
Molecular Research Center DNAZOL 50ML
SDP
Catalog No. NC0304199
Encompass
Genomic DNA isolation reagent for use with tissues, cells and liquids. The isolation is fast (10-30 min), simple and reliable. Non-toxic. 1 ml of DNAzol isolates DNA from 25-50 mg tissue or 1-3 x 107 cells.
Catalog No. NC0304199 Supplier Molecular Research Center Supplier No. DN 12750ML
Genomic DNA isolation reagent for use with tissues, cells and liquids. The isolation is fast (10-30 min), simple and reliable. Non-toxic. 1 ml of DNAzol isolates DNA from 25-50 mg tissue or 1-3 x 107 cells.