Learn More
Molecular Research Center DNAzol Direct, 25 mL
SDP
Catalog No. NC1213875
Encompass
DNAzol Direct is a universal reagent for processing biological samples for direct PCR. No DNA isolation is required. DNAzol Direct is designed to process a wide range of samples including animal, plant, fungi, yeast, bacterial, and viral samples. It has been used with blood, serum and plasma, saliva, buccal swabs, blood cards, hairs and feathers, leaves and seeds, and formalin-fixed tissues.
Catalog No. NC1213875 Supplier Molecular Research Center Supplier No. DN 13125ML
DNAzol Direct is a universal reagent for processing biological samples for direct PCR. No DNA isolation is required. DNAzol Direct is designed to process a wide range of samples including animal, plant, fungi, yeast, bacterial, and viral samples. It has been used with blood, serum and plasma, saliva, buccal swabs, blood cards, hairs and feathers, leaves and seeds, and formalin-fixed tissues.