Learn More
Molecular Research Center DNAZOL DIRECT
SDP
Catalog No. NC0109325
Encompass
A universal reagent for sample processing for direct PCR. Lyse sample in DNAzol Direct, incubate and use lysate as PCR template. No DNA isolation necessary. Non-toxic. Use 50 ul reagent per 5 ul whole blood or 1 mg tissue.
Catalog No. NC0109325 Supplier Molecular Research Center Supplier No. DN 131
A universal reagent for sample processing for direct PCR. Lyse sample in DNAzol Direct, incubate and use lysate as PCR template. No DNA isolation necessary. Non-toxic. Use 50 ul reagent per 5 ul whole blood or 1 mg tissue.