Learn More
Thermo Scientific Chemicals DNAzol™ Reagent
Description
DNAzol™ Reagent is used as an electrophoretic reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Cholesterol (cholesterin, cholesteryl alcohol) is a lipid sterol found in vertebrate cell membranes
- Cholesterol is a cholestanoid, a class of molecule that contains the C-27 cholestane motif
- A precursor of many steroids and sex hormones in animals including cortisol, aldosterone, testosterone, progesterone, and estrogen
- It plays a role in neurotransmission and is crucial for organ systems, such as immune, reproductive and endocrine systems
Applications
- Suitable for in vitro and in vivo applications
- Stabilized in powder form for laboratory use only
Specifications
Specifications
| Color | Blue-green |
| For Use With (Application) | Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin |
| Product Type | DNA Extraction Reagent |
| Quantity | 50 mL |
| Form | Liquid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.