Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DNDS (disodium) | 3709-43-1 | 99.4% | 5 G
SDP

Catalog No. 5000342351
Encompass_Preferred

DNDS (disodium) is a chemical substance primarily used as an erythrocyte sedimentation inhibitor in laboratory applications and for the manufacture of other substances. This compound has a molecular weight of 474.33, a chemical formula of C14H8N2Na2O10S2, and appears as a light yellow to yellow solid. It is soluble in water at 83.33 mg/mL (175.68 mM) and is stable under recommended storage conditions.

  • Potent erythrocyte sedimentation inhibitor
  • Purity of 99.4%
  • CAS number: 3709-43-1
  • Molecular weight: 474.33
  • Chemical formula: C14H8N2Na2O10S2
  • Solid appearance, light yellow to yellow color
  • Boiling point of 552 °C at 760 mmHg
  • Stable under recommended storage conditions
  • Storage at 4°C, sealed, away from moisture and light
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
  • Soluble in H2O at 83.33 mg/mL (175.68 mM)

Catalog No. 50-003-42351 Supplier Medchemexpress LLC Supplier No. HY1295605G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNDS (disodium) is a chemical substance primarily used as an erythrocyte sedimentation inhibitor in laboratory applications and for the manufacture of other substances. This compound has a molecular weight of 474.33, a chemical formula of C14H8N2Na2O10S2, and appears as a light yellow to yellow solid. It is soluble in water at 83.33 mg/mL (175.68 mM) and is stable under recommended storage conditions.

  • Potent erythrocyte sedimentation inhibitor
  • Purity of 99.4%
  • CAS number: 3709-43-1
  • Molecular weight: 474.33
  • Chemical formula: C14H8N2Na2O10S2
  • Solid appearance, light yellow to yellow color
  • Boiling point of 552 °C at 760 mmHg
  • Stable under recommended storage conditions
  • Storage at 4°C, sealed, away from moisture and light
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
  • Soluble in H2O at 83.33 mg/mL (175.68 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.