Learn More
Aviva Systems Biology Corp DNER Antibody - middle region (ARP50777_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50777_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DNER. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DNER. Alternate names: UNQ26, betApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60431