Learn More
AdipoGen DNER ED hFc h rec
SDP
Catalog No. 502562988
Encompass_Preferred
Protein. The extracellular domain of human DNER aa 29-637 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.06EU/µg purified protein LAL test Lonza. Lyophilized from 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. DNER is an activator of the Notch1 pathway. It mediates neuron-glia interaction during astrocytogenesis.
Catalog No. 50-256-2988 Supplier AdipoGen Supplier No. AG40A0119TC010
Protein. The extracellular domain of human DNER aa 29-637 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.06EU/µg purified protein LAL test Lonza. Lyophilized from 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. DNER is an activator of the Notch1 pathway. It mediates neuron-glia interaction during astrocytogenesis.