Learn More
AdipoGen DNER ED msFc h rec
SDP
Catalog No. 502562959
Encompass_Preferred
Protein. Signal peptide and extracellular domain of mouse DNER aa 1-640 are fused at the C-terminus to the Fc portion of human IgG. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >85% SDS-PAGE. DNER is an activator of the Notch1 pathway. It mediates neuron-glia interaction during astrocytogenesis.
Catalog No. 50-256-2959 Supplier AdipoGen Supplier No. AG40A0177C010
Protein. Signal peptide and extracellular domain of mouse DNER aa 1-640 are fused at the C-terminus to the Fc portion of human IgG. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >85% SDS-PAGE. DNER is an activator of the Notch1 pathway. It mediates neuron-glia interaction during astrocytogenesis.