Learn More
Aviva Systems Biology Corp DNM1L Antibody - N-terminal region (ARP52329_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52329_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNM1L. It was validated on Western Blot. DNM1L,dynamin 1-like,DLP1; DRP1; DVLP; VPS1; DYMPLE; HDYNIV; DYNIV-11; FLJ41912; DNM1L,dynamin-1-like protein,dynamin-1-like protein,dynamin-like protein 4,dynamin-like protein IV,Dnm1p/Vps1p-like protein,dynamin-related protein 1,dynamin family member proline-rich carboxyl-terminal domain lessThe immunogen is a synthetic peptide directed towards the N terminal region of human DNM1L. Alternate names: DLP1, DRP1, DVLP, DYMPLE, HDYNIV, VPS1, EMPF, DYNIV-11ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-707