Learn More
Aviva Systems Biology Corp Dnm3 Antibody - C-terminal region (ARP55282_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55282_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnm3. It was validated on Western Blot. . Alternate names: 9630020E24Rik, AW061159, B230343F03Rik, C530045C17, mKIAA0820ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-024