Learn More
Aviva Systems Biology Corp Dnmt1 Antibody - N-terminal region (ARP37033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnmt1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of mouse Dnmt1. Alternate names: Cxxc9, Dnmt, Dnmt1o, MTase, Met-1, Met1, MommeD2, MCMT, m.MmuIApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-568