Learn More
Enzo Life Sciences Dnmt1 monoclonal antibody (60B1220.1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804369C100
Encompass_Preferred
Clone: 60B1220.1. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 637-650 of human Dnmt1 (DNA methyltransferase-1). Species reactivity: Human, Mouse Zebrafish. Applications: ICC, IHC (PS), IP, WB ChIP. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.05% BSA and 0.05% sodium azide.
Catalog No. 50-200-9892