Learn More
Epigentek Inc DNMT1 Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A1700100
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Protein of Human TET2. Tested Applications: WB, IHC. Reactivity: Human, Mouse, Rat. Alternative Names: MDS, KIAA1546.
Catalog No. 50-109-4982