Learn More
Sino Biological Recombinant Human COMT Protein (GST Tag), Active
Catalog No. 502575573
Encompass_Preferred
Recombinant human COMT (27-271) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5573 Supplier Sino Biological Supplier No. C339381G50
Recombinant human COMT (27-271) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.