Learn More
Medchemexpress LLC DNMT3A-IN-1, 50mg
SDP
Catalog No. NC2226264
Encompass_Preferred
DNMT3A-IN-1 is a potent and selective DNMT3A inhibitor. DNMT3A-IN-1 shows inhibitor activities against DNMT3A with kI values range from 9.16-18.85 μM (AdoMet) and 11.37-23.34 μM (poly dI-dC), 1403598-56-0, 50mg
This item is not returnable. View return policy
Catalog No. NC2226264 Supplier Medchemexpress LLC Supplier No. HY14443350MG
DNMT3A-IN-1 is a potent and selective DNMT3A inhibitor. DNMT3A-IN-1 shows inhibitor activities against DNMT3A with kI values range from 9.16-18.85 μM (AdoMet) and 11.37-23.34 μM (poly dI-dC), 1403598-56-0, 50mg