Learn More
Cayman Chemical Danofloxacin (mesylate), Purity: ≥98%, CAS: 119478-55-6, 100 mg
SDP
Catalog No. 502986471
Encompass_Preferred
A fluroquinolone antibiotic; has activity against 68 field isolates of Mycoplasma species isolated from cattle, swine, and poultry (MICs = 8-500 nM); protective in vivo against P. multocida, E. coli, and S. choleraesuis in mice with protective doses of 0.31, 0.40, and 2.42 mg/kg, respectively
Catalog No. 50-298-6471 Supplier Cayman Chemical Supplier No. 22408100 MG
A fluroquinolone antibiotic; has activity against 68 field isolates of Mycoplasma species isolated from cattle, swine, and poultry (MICs = 8-500 nM); protective in vivo against P. multocida, E. coli, and S. choleraesuis in mice with protective doses of 0.31, 0.40, and 2.42 mg/kg, respectively