Learn More
Cayman Chemical Dnotefurn 50mg
SDP
Catalog No. 502946959
Encompass_Preferred
A neonicotinoid insecticide; a nAChR agonist (EC50 = 7.6 µM for the induction of inward currents in isolated P. americana neurons); induces mortality in adult P. americana; lethal to insecticide-sensitive or -resistant strains of A. gambiae, C. quinquefasciatus, and A. aegypti in larval (LC50s = 0.14-0.44 mg/L) and adult (LD50s = 0.18-31.16 ng/mg) bioassays; has been found in a variety of commercial foodstuffs, including rice, cucumber, milk, egg, and pork samples, as well as in water samples from freshwater streams
Catalog No. 50-294-6959 Supplier Cayman Chemical Supplier No. 3380250 MG
A neonicotinoid insecticide; a nAChR agonist (EC50 = 7.6 µM for the induction of inward currents in isolated P. americana neurons); induces mortality in adult P. americana; lethal to insecticide-sensitive or -resistant strains of A. gambiae, C. quinquefasciatus, and A. aegypti in larval (LC50s = 0.14-0.44 mg/L) and adult (LD50s = 0.18-31.16 ng/mg) bioassays; has been found in a variety of commercial foodstuffs, including rice, cucumber, milk, egg, and pork samples, as well as in water samples from freshwater streams