Learn More
Medchemexpress LLC DNP-PEG3-azide | 951671-87-7 | 99.2% | C14H20N6O7 | 25 MG
SDP
Catalog No. 5000352677
Encompass_Preferred
DNP-PEG3-azide is a PEG-based PROTAC linker crucial for the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an azide group that enables both copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. It is a light yellow to yellow viscous liquid with a high purity of 99.18%.
Catalog No. 50-003-52677 Supplier Medchemexpress LLC Supplier No. 25MGHY140617
DNP-PEG3-azide is a PEG-based PROTAC linker crucial for the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an azide group that enables both copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. It is a light yellow to yellow viscous liquid with a high purity of 99.18%.