Learn More
Medchemexpress LLC Dnp-peg4-acid | 858126-76-8 | 1 G
SDP
Catalog No. 5000341000
Encompass_Preferred
DNP-PEG4-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds featuring two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting a specific protein. These compounds utilize the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Peg-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • Appearance: viscous liquid
  • Color: colorless to light yellow
Catalog No. 50-003-41000 Supplier Medchemexpress LLC Supplier No. HY1301981G
DNP-PEG4-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds featuring two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting a specific protein. These compounds utilize the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Peg-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • Appearance: viscous liquid
  • Color: colorless to light yellow