Learn More
Medchemexpress LLC DNP-PEG4-acid | 858126-76-8 | 100 MG
SDP
Catalog No. 5000338565
Encompass_Preferred
DNP-PEG4-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, consisting of two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Relevant for cancer and cancer targeted therapy research
Catalog No. 50-003-38565 Supplier Medchemexpress LLC Supplier No. HY130198100MG
DNP-PEG4-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, consisting of two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Relevant for cancer and cancer targeted therapy research