Learn More
Medchemexpress LLC DNP-PEG4-acid | 858126-76-8 | >98% | 250 MG
SDP
Catalog No. 5000341487
Encompass_Preferred
DNP-PEG4-acid is a PEG-based PROTAC linker for the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for research use only.
  • PEG-based PROTAC linker
  • Enables the synthesis of PROTACs
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-41487 Supplier Medchemexpress LLC Supplier No. 250MGHY130198
DNP-PEG4-acid is a PEG-based PROTAC linker for the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for research use only.
  • PEG-based PROTAC linker
  • Enables the synthesis of PROTACs
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
  • For research use only