Learn More
Medchemexpress LLC DNP-PEG4-DBCO | 2149585-17-9 | 97.0% | C35H39N5O10 | 50 MG
SDP
Catalog No. 5000353229
Encompass_Preferred
DNP-PEG4-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide-containing molecules
Catalog No. 50-003-53229 Supplier Medchemexpress LLC Supplier No. HY14061950MG
DNP-PEG4-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide-containing molecules