Learn More
Medchemexpress LLC Dnp-peg4-dbco | 2149585-17-9 | MFCD31654037 | 97.0% | C35H39N5O10 | 10MG
SDP
Catalog No. 5000259698
Encompass_Preferred
DNP-PEG4-DBCO is a PEG-based PROTAC linker and click-chemistry reagent that combines a dinitrophenyl (DNP) moiety with a dibenzocyclooctyne (DBCO) group. The DBCO enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules, supporting efficient bioconjugation and PROTAC assembly.
  • Contains DNP and DBCO functional groups for dual reactivity.
  • Enables copper-free SPAAC click chemistry with azide partners.
  • Suitable for PROTAC linker synthesis and bioconjugation workflows.
  • High purity (97.0%) for reliable analytical and preparative use.
  • Stable under recommended storage conditions for extended shelf life.
Catalog No. 50-002-59698 Supplier Medchemexpress LLC Supplier No. HY14061910MG
DNP-PEG4-DBCO is a PEG-based PROTAC linker and click-chemistry reagent that combines a dinitrophenyl (DNP) moiety with a dibenzocyclooctyne (DBCO) group. The DBCO enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules, supporting efficient bioconjugation and PROTAC assembly.
  • Contains DNP and DBCO functional groups for dual reactivity.
  • Enables copper-free SPAAC click chemistry with azide partners.
  • Suitable for PROTAC linker synthesis and bioconjugation workflows.
  • High purity (97.0%) for reliable analytical and preparative use.
  • Stable under recommended storage conditions for extended shelf life.