Learn More
Medchemexpress LLC DNP-PEG4-NHS ester | 858126-78-0 | 96.8% | C21H28N4O12 | 250 MG
SDP
Catalog No. 5000352739
Encompass_Preferred
DNP-PEG4-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins, functioning through two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other binding to the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system to degrade target proteins
  • Viscous liquid appearance
  • Light yellow to green yellow color
  • Soluble in DMSO (≥ 100 mg/mL)
  • Insoluble in H2O (< 0.1 mg/mL)
Catalog No. 50-003-52739 Supplier Medchemexpress LLC Supplier No. 250MGHY140614
DNP-PEG4-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins, functioning through two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other binding to the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system to degrade target proteins
  • Viscous liquid appearance
  • Light yellow to green yellow color
  • Soluble in DMSO (≥ 100 mg/mL)
  • Insoluble in H2O (< 0.1 mg/mL)