Learn More
LABELS DIRECT INC 3.26" X 984' (300M) - DNP R510HF Ultra Durable Resin - Zebra Industrial (CSO) Printer Ribbons - 24/Case - Black
SDP
Catalog No. NC3628058
Encompass
3.26" X 984' (300M) - R510HF Ultra Durable Resin, DNP Thermal Transfer Ribbons, Zebra Industrial Printers, Coated Side Out (CSO) Ribbons, 24/Case, 1" Fiber Core, Black
Catalog No. NC3628058 Supplier LABELS DIRECT INC Supplier No. 18105289
3.26" X 984' (300M) - R510HF Ultra Durable Resin, DNP Thermal Transfer Ribbons, Zebra Industrial Printers, Coated Side Out (CSO) Ribbons, 24/Case, 1" Fiber Core, Black