Learn More
Biotium DNP Tyramide
Catalog No. 501964997
Encompass_Preferred
DNP tyramide conjugates can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ.
Catalog No. 50-196-4997 Supplier Biotium Supplier No. 96019
DNP tyramide conjugates can be used for tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ.