Learn More
Medchemexpress LLC DNQX | 2379-57-9 | C8H4N4O6 | 50 MG
SDP
Catalog No. 5000375119
Encompass_Preferred
DNQX (FG 9041) is a quinoxaline derivative that acts as a selective, potent competitive non-NMDA glutamate receptor antagonist. It exhibits IC50 values of 0.5 μM for AMPA receptors, 2 μM for kainate receptors, and 40 μM for NMDA receptors.
  • Selective non-NMDA glutamate receptor antagonist
  • Potent competitive antagonist
  • IC50s: 0.5 μM (AMPA), 2 μM (kainate), 40 μM (NMDA)
  • High purity (99.67%)
  • Available as solid and solution
  • In vivo dissolution protocols provided
  • In vitro, depolarizes thalamic reticular nucleus (TRN) neurons
  • In vivo, diminishes phencyclidine (PCP) hsp70 induction
Catalog No. 50-003-75119 Supplier Medchemexpress LLC Supplier No. HY1506750MG
DNQX (FG 9041) is a quinoxaline derivative that acts as a selective, potent competitive non-NMDA glutamate receptor antagonist. It exhibits IC50 values of 0.5 μM for AMPA receptors, 2 μM for kainate receptors, and 40 μM for NMDA receptors.
  • Selective non-NMDA glutamate receptor antagonist
  • Potent competitive antagonist
  • IC50s: 0.5 μM (AMPA), 2 μM (kainate), 40 μM (NMDA)
  • High purity (99.67%)
  • Available as solid and solution
  • In vivo dissolution protocols provided
  • In vitro, depolarizes thalamic reticular nucleus (TRN) neurons
  • In vivo, diminishes phencyclidine (PCP) hsp70 induction