Learn More
Medchemexpress LLC HY-15067 25mg Medchemexpress, DNQX CAS:2379-57-9 Purity:>98%
SDP
Catalog No. 502081678
Encompass_Preferred
Medchemexpress, HY-15067 25mg DNQX CAS:2379-57-9 DNQX (FG 9041), a quinoxaline derivative, is a selective, potent competitive non-NMDA glutamate receptor antagonist (IC50s = 0.5, 2 and 40 μM for AMPA, kainate and NMDA receptors, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1678 Supplier Medchemexpress LLC Supplier No. HY1506725MG
Medchemexpress, HY-15067 25mg DNQX CAS:2379-57-9 DNQX (FG 9041), a quinoxaline derivative, is a selective, potent competitive non-NMDA glutamate receptor antagonist (IC50s = 0.5, 2 and 40 μM for AMPA, kainate and NMDA receptors, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.