Learn More
Selleck Chemical LLC DNQX-E1259-25MG
SDP
Catalog No. 503138655
Encompass_Preferred
DNQX is an AMPA receptor antagonist that prevents excess calcium influx via voltage-gated calcium channels
Catalog No. 50-313-8655 Supplier Selleck Chemical LLC Supplier No. E125925MG
DNQX is an AMPA receptor antagonist that prevents excess calcium influx via voltage-gated calcium channels