Learn More
Selleck Chemical LLC DNQX-E1259-5MG
SDP
Catalog No. 503138653
Encompass_Preferred
DNQX is an AMPA receptor antagonist that prevents excess calcium influx via voltage-gated calcium channels
Catalog No. 50-313-8653 Supplier Selleck Chemical LLC Supplier No. E12595MG
DNQX is an AMPA receptor antagonist that prevents excess calcium influx via voltage-gated calcium channels