Learn More
Medchemexpress LLC Dnqx (FG 9041) | 2379-57-9 | C8H4N4O6 | 10 MM 1 ML
SDP
Catalog No. 5000375280
Encompass_Preferred
DNQX (FG 9041) is a quinoxaline derivative that functions as a selective, potent competitive non-NMDA glutamate receptor antagonist, demonstrating IC50 values of 0.5, 2, and 40 μM for AMPA, kainate, and NMDA receptors, respectively.
  • Selectively depolarizes thalamic reticular nucleus (TRN) neurons in vitro.
  • Significantly diminishes phencyclidine (PCP) induced hsp70 in posterior cingulate and retrosplenial cortex in vivo.
Catalog No. 50-003-75280 Supplier Medchemexpress LLC Supplier No. HY1506710MM/1ML
DNQX (FG 9041) is a quinoxaline derivative that functions as a selective, potent competitive non-NMDA glutamate receptor antagonist, demonstrating IC50 values of 0.5, 2, and 40 μM for AMPA, kainate, and NMDA receptors, respectively.
  • Selectively depolarizes thalamic reticular nucleus (TRN) neurons in vitro.
  • Significantly diminishes phencyclidine (PCP) induced hsp70 in posterior cingulate and retrosplenial cortex in vivo.