Learn More
Medchemexpress LLC DNQX (FG 9041) | 2379-57-9 | C8H4N4O6 | 200 MG
SDP
Catalog No. 5000375118
Encompass_Preferred
DNQX (FG 9041) is a quinoxaline derivative and a selective, potent competitive non-NMDA glutamate receptor antagonist. It is characterized by its activity against various glutamate receptors.
  • Selective, potent competitive non-NMDA glutamate receptor antagonist
  • Targets AMPA receptors with an IC50 of 0.5 μM
  • Targets kainate receptors with an IC50 of 2 μM
  • Targets NMDA receptors with an IC50 of 40 μM
  • Selectively depolarizes thalamic reticular nucleus (TRN) neurons in vitro
  • Diminishes phencyclidine (PCP) induced hsp70 induction in vivo
Catalog No. 50-003-75118 Supplier Medchemexpress LLC Supplier No. HY15067200MG
DNQX (FG 9041) is a quinoxaline derivative and a selective, potent competitive non-NMDA glutamate receptor antagonist. It is characterized by its activity against various glutamate receptors.
  • Selective, potent competitive non-NMDA glutamate receptor antagonist
  • Targets AMPA receptors with an IC50 of 0.5 μM
  • Targets kainate receptors with an IC50 of 2 μM
  • Targets NMDA receptors with an IC50 of 40 μM
  • Selectively depolarizes thalamic reticular nucleus (TRN) neurons in vitro
  • Diminishes phencyclidine (PCP) induced hsp70 induction in vivo