Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dnsah-13C6 | MFCD00129753 | 100.0% | 248.10 g/mol | C13C6H6N4O6 | 1 MG
SDP

Catalog No. 5000282085
Encompass_Preferred

DNSAH-13C6 is a carbon-13 (13C6) stable isotope-labeled analogue of DNSAH supplied as a solid for use as a tracer or internal standard in quantitative analyses, including NMR, GC-MS, and LC-MS. For research use only.

  • Stable isotope-labeled tracer for metabolic flux and quantitative analysis.
  • Chemical purity 99.98%.
  • Isotopic labeling: carbon-13 (13C6).
  • Molecular formula C13C6H6N4O6; molecular weight 248.10 g/mol.
  • Light yellow to yellow solid form.
  • Storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • Intended for use as a tracer or internal standard in NMR, GC-MS, and LC-MS.

Catalog No. 50-002-82085 Supplier Medchemexpress LLC Supplier No. HYW280531S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNSAH-13C6 is a carbon-13 (13C6) stable isotope-labeled analogue of DNSAH supplied as a solid for use as a tracer or internal standard in quantitative analyses, including NMR, GC-MS, and LC-MS. For research use only.

  • Stable isotope-labeled tracer for metabolic flux and quantitative analysis.
  • Chemical purity 99.98%.
  • Isotopic labeling: carbon-13 (13C6).
  • Molecular formula C13C6H6N4O6; molecular weight 248.10 g/mol.
  • Light yellow to yellow solid form.
  • Storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • Intended for use as a tracer or internal standard in NMR, GC-MS, and LC-MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.