Learn More
Cayman Chemical Dansyl Chloride, Purity: ≥95%, CAS: 605-65-2, 25 g
SDP
Catalog No. 502937224
Encompass_Preferred
A reactive probe for derivatization of primary amines, including those on amino acids, peptides, and polyamines, for detection by HPLC; has been used for the derivatization of phenol-containing compounds, such as steroids, for detection by MS/MS; a fluorescent probe for proteins and enzymes with ex/em = 340/535 nm, respectively, in acetone
Catalog No. 50-293-7224 Supplier Cayman Chemical Supplier No. 2745825 G
A reactive probe for derivatization of primary amines, including those on amino acids, peptides, and polyamines, for detection by HPLC; has been used for the derivatization of phenol-containing compounds, such as steroids, for detection by MS/MS; a fluorescent probe for proteins and enzymes with ex/em = 340/535 nm, respectively, in acetone